Learn More
PDC Direct Thermal Paper Label Permanent, 760/Roll, 8 Rolls/Case
Catalog No. 21235857
Change view
Click to view available options
Quantity:
760/Roll, 8 Rolls/Cs.
Catalog No. Quantity
21-235-857 760/Roll, 8 Rolls/Cs.
1 options
Catalog No. 21-235-857 Supplier PDC Supplier No. THERMD25
Add to Cart
Add to Cart
Designed for Eltron LP and TLP printers, these labels ensure strong adhesion, clear printing, and versatile performance for labeling tasks.
  • Designed for Eltron LP, TLP Series, and other widely used printers to support seamless use
  • Permanent adhesive provides strong and durable labeling that withstands daily handling
  • Direct thermal printing technology removes the need for ink or ribbons, reducing maintenance
  • Ideal for versatile applications across healthcare, inventory, asset tracking, and general labeling
  • Rounded corner design helps prevent peeling and curling, ensuring smooth application
  • Perforated layout makes label separation quick and efficient for faster workflows
  • Optimized roll design ensures compatibility with standard thermal printers
  • Glove-friendly adhesive enhances usability in medical, laboratory, and industrial settings
  • Bulk packaging offers continuous supply for high-demand labeling environments
  • Trusted multi-purpose labeling solution that delivers consistent quality and performance
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 760/Roll, 8 Rolls/Cs.
Dimensions (L x W) 3 1/2 x 1 in.
Length (English) 3.5 in.
Width (English) 1 in.
Width (Metric) 25.4 mm