Learn More
PDC Direct Thermal Paper Label Permanent, 960/Roll, 8 Rolls/Case
Catalog No. 21235850
Change view
Click to view available options
Quantity:
960/Roll, 8 Rolls/Cs.
Catalog No. Quantity
21-235-850 960/Roll, 8 Rolls/Cs.
1 options
Catalog No. 21-235-850 Supplier PDC Supplier No. THERMD17
Add to Cart
Add to Cart
Designed for clear printing and strong adhesion, ensuring reliable performance across applications.
  • Permanent adhesive for secure, long-lasting label placement on different surfaces.
  • Ideal for versatile applications across a wide range of industries.
  • Compatible with multiple thermal printers for smooth integration.
  • Direct thermal printing provides sharp, ribbon-free images with reduced maintenance.
  • White paper material enhances print contrast and ensures clarity for readability.
  • Designed to minimize print jams and streamline workflow efficiency.
  • Suitable for healthcare, logistics, and general labeling tasks.
  • Convenient multi-roll pack ensures continuous supply and productivity.
  • Trusted for consistent quality and reliable performance in demanding environments.
  • Glove-friendly adhesive allows easy application in medical and industrial settings.
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 960/Roll, 8 Rolls/Cs.
Dimensions (L x W) 2 x 1 1/2 in.
Length (English) 2 in.
Width (English) 1.5 in.
Width (Metric) 38.1 mm