Learn More
PDC Direct Thermal Paper Label Permanent, 960/Roll, 8 Rolls/Case
Catalog No. 21235854
Change view
Click to view available options
Quantity:
960/Roll, 8 Rolls/Cs.
Catalog No. Quantity
21-235-854 960/Roll, 8 Rolls/Cs.
1 options
Catalog No. 21-235-854 Supplier PDC Supplier No. THERMD20
Add to Cart
Add to Cart
Compatible with Eltron LP and TLP printers, these labels ensure reliable adhesion, clear printing, and versatile multi-purpose use.
  • Compatible with Eltron LP, TLP Series, and other widely used printers
  • Permanent adhesive ensures strong and reliable label application on various surfaces
  • Direct thermal printing requires no ink or ribbon, simplifying workflow and reducing costs
  • Designed for multiple labeling needs across healthcare, asset tracking, and inventory management
  • Rounded corner design helps prevent peeling and curling, maintaining secure placement
  • Perforated layout allows smooth and easy label separation for faster operations
  • Optimized roll format fits standard printer setups for consistent performance
  • Glove-friendly adhesive makes application easier in medical, lab, and industrial environments
  • Bulk packaging provides consistent supply to support high-volume labeling tasks
  • Multi-purpose functionality supports specimen labeling, asset management, and general tracking
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 960/Roll, 8 Rolls/Cs.
Dimensions (L x W) 2 1/2 x 1 1/2 in.
Length (English) 2.5 in.
Width (English) 1.5 in.
Width (Metric) 38.1 mm