Learn More
PDC Direct Thermal Paper Label Permanent, 960/Roll, 8 Rolls/Case
Catalog No. 21235858
Change view
Click to view available options
Quantity:
960/Roll, 8 Rolls/Cs.
Catalog No. Quantity
21-235-858 960/Roll, 8 Rolls/Cs.
1 options
Catalog No. 21-235-858 Supplier PDC Supplier No. THERMD26
Add to Cart
Add to Cart
Compatible with major printers, featuring strong adhesive and bulk packaging for versatile, multi-purpose applications.
  • Designed for printers including Eltron LP and TLP series
  • Permanent adhesive ensures long-lasting label application
  • Direct thermal printing eliminates the need for ink or ribbons
  • Rounded corners prevent label peeling and curling
  • Perforated design allows smooth, easy label separation
  • Roll design fits most printers for reliable printing
  • Glove-friendly adhesive suitable for healthcare and lab settings
  • Bulk supply supports high-volume labeling needs
  • Ideal for specimen labeling, asset tagging, and inventory management
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 960/Roll, 8 Rolls/Cs.
Dimensions (L x W) 4 x 1 1/2 in.
Length (English) 4 in.
Width (English) 1.5 in.
Width (Metric) 38.1 mm