Learn More
PDC Direct Thermal Paper Label, White with Orange, 1000 Labels/Roll
Catalog No. 21235791
Change view
Click to view available options
Quantity:
1000 Labels/Roll
Catalog No. Quantity
21-235-791 1000 Labels/Roll
1 options
Catalog No. 21-235-791 Supplier PDC Supplier No. LDTSLTO1
Add to Cart
Add to Cart
White labels with orange highlights provide durable adhesion and high visibility, making them ideal for demanding labeling applications.
  • Large-format design supports clear, detailed labeling in clinical and industrial settings
  • White background with orange accents enhances visibility for quick identification
  • Permanent adhesive ensures secure placement on containers and packages
  • Compatible with bulk and industrial dispensers for smooth use
  • Reliable performance for healthcare, logistics, and inventory workflows
  • High-volume supply helps maintain labeling efficiency
Specifications
Adhesive Type Permanent
Coating None
Color White, Orange
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 1000 Labels/Roll
Dimensions (L x W) 4 x 5 1/2 in.
Length (English) 4 in.
Width (English) 5.5 in.
Width (Metric) 139.7 mm