Learn More
PDC Direct Thermal Paper Label, Yellow, 280 Labels/Roll, 36 Rolls/Case
Catalog No. 21235790
Change view
Click to view available options
Quantity:
280 Labels/Roll, 36 Rolls/Cs.
Catalog No. Quantity
21-235-790 280 Labels/Roll, 36 Rolls/Cs.
1 options
Catalog No. 21-235-790 Supplier PDC Supplier No. LDQLPY320
Add to Cart
Add to Cart
High-visibility yellow direct thermal labels with permanent adhesive and black sensor bar for reliable specimen and patient identification.
  • Bright yellow labels enhance visibility for critical specimen and patient identification
  • Permanent adhesive ensures labels remain securely affixed to tubes, vials, and containers
  • Black sensor bar enables accurate alignment and smooth printer operation
  • Durable paper stock provides sharp, legible printing for healthcare and lab workflows
  • Supports efficient specimen tracking, reducing labeling errors and delays
  • Bulk supply packaging ensures cost-effective use in clinical and laboratory settings
Specifications
Adhesive Type Permanent
Coating None
Color Yellow
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 280 Labels/Roll, 36 Rolls/Cs.
Dimensions (L x W) 2 x 1 1/4 in.
Length (English) 2 in.
Width (English) 1.25 in.
Width (Metric) 31.75 mm