Learn More
PDC Direct Thermal Paper Labels, White, 1135/Roll, 12 Rolls/Box
Catalog No. 21235818
Change view
Click to view available options
Quantity:
1135/Roll, 12 Rolls/Box
Catalog No. Quantity
21-235-818 1135/Roll, 12 Rolls/Box
1 options
Catalog No. 21-235-818 Supplier PDC Supplier No. TD1114214
  • $214.00
    $159.65 / Case of 12
    Save $54.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Deliver smudge-free printing with strong adhesion, ensuring dependable performance in healthcare, lab, and clinical use.
  • White direct thermal paper labels with crisp print quality for clear identification
  • Label size: 2 1/4 in. x 1 1/4 in. for versatile labeling needs
  • Supplied 1,135 labels per roll and 12 rolls per box for extended supply
  • Permanent adhesive ensures secure placement on a variety of surfaces
  • 1 in. core compatible with most standard direct thermal printers
  • Ribbonless design supports fast and efficient printing operations
  • Reliable solution for healthcare, clinical, and laboratory labeling tasks
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 1135/Roll, 12 Rolls/Box
Dimensions (L x W) 2 1/4 x 1 1/4 in.
Length (English) 2.25 in.
Width (English) 1.25 in.
Width (Metric) 31.75 mm