Learn More
PDC Direct Thermal Paper Permanent Label, 2900 Labels/Roll, 3 Rolls/Box
Catalog No. 21235826
Change view
Click to view available options
Quantity:
2900 Labels/Roll, 3 Rolls/Box
Catalog No. Quantity
21-235-826 2900 Labels/Roll, 3 Rolls/Box
1 options
Catalog No. 21-235-826 Supplier PDC Supplier No. TD24
  • $336.00
    $251.65 / Case of 3
    Save $84.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Durable direct thermal labels with permanent adhesive, perfect for everyday healthcare, logistics, and inventory labeling needs.
  • Direct thermal printable technology provides crisp, clear labels without the need for ink or ribbons
  • Permanent adhesive ensures labels stay firmly in place on various surfaces
  • High-quality white label surface enhances readability and scanning accuracy
  • Durable material suitable for frequent and heavy-duty use
  • Easy to handle and apply for improved operational efficiency
  • Designed for reliable performance in healthcare, logistics, and inventory environments
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 2900 Labels/Roll, 3 Rolls/Box
Dimensions (L x W) 4 x 2 in.
Length (English) 4 in.
Width (English) 2 in.
Width (Metric) 50.8 mm