Learn More
PDC Direct Thermal Slit Label, 200 Labels/Roll
Catalog No. 21235835
Change view
Click to view available options
Quantity:
200 Labels/Roll
Catalog No. Quantity
21-235-835 200 Labels/Roll
1 options
Catalog No. 21-235-835 Supplier PDC Supplier No. TD7522316
Add to Cart
Add to Cart
Durable direct thermal slit labels with permanent adhesive, designed for reliable printing and scanning with enhanced print compatibility.
  • Direct thermal slit label with permanent adhesive
  • White color for clear printing and readability
  • Black sensor bar ensures accurate printer detection and alignment
  • Reliable and consistent label performance for scanning and identification
  • Suitable for healthcare, lab, and other professional labeling needs
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 200 Labels/Roll
Dimensions (L x W) 2 x 2 3/16 in.
Length (English) 2 in.
Width (English) 2.1875 in.
Width (Metric) 55.5625 mm