Learn More
PDC Direct Thermal Synthetic Label, 2000 Labels/Roll
Catalog No. 21235800
Change view
Click to view available options
Quantity:
2000 Labels/Roll
Catalog No. Quantity
21-235-800 2000 Labels/Roll
1 options
Catalog No. 21-235-800 Supplier PDC Supplier No. LDWHP173
Add to Cart
Add to Cart
Durable synthetic labels with permanent adhesive, designed for smooth integration with Epic EHR systems in healthcare and lab settings.
  • Compatible with Epic EHR systems including Beaker and Lab modules for seamless workflow
  • Synthetic construction resists moisture, smudging, and handling in clinical environments
  • White face stock supports sharp, clear printing for accurate identification
  • Permanent adhesive secures labels firmly on samples, vials, and containers
  • 1 in. core fits compact dispensers for easy handling in busy labs
  • Each roll provides 2000 labels to support moderate-volume labeling needs
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Synthetic
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 2000 Labels/Roll
Dimensions (L x W) 2 x 1 1/4 in.
Length (English) 2 in.
Width (English) 1.25 in.
Width (Metric) 31.75 mm