Learn More
PDC Direct Thermal Synthetic Label, 5000 Labels/Roll
Catalog No. 21235797
Change view
Click to view available options
Quantity:
5000 Labels/Roll
Catalog No. Quantity
21-235-797 5000 Labels/Roll
1 options
Catalog No. 21-235-797 Supplier PDC Supplier No. LDWHP148
Add to Cart
Add to Cart
Durable, chemical-resistant synthetic labels with permanent adhesive, designed for reliable use in Epic and Cerner EHR healthcare systems.
  • Compatible with Epic and Cerner EHR platforms, including Beaker and Lab modules
  • Synthetic material resists alcohol, water, and other chemicals for dependable performance
  • White face stock supports sharp, clear printing for accurate identification
  • Permanent adhesive secures labels firmly to specimens, vials, and lab containers
  • 3 in. core fits standard dispensers for smooth, high-volume label handling
  • Each roll includes 5000 labels, supporting large-scale clinical and laboratory workflows
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Synthetic
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 5000 Labels/Roll
Dimensions (L x W) 2 x 1 1/4 in.
Length (English) 2 in.
Width (English) 1.25 in.
Width (Metric) 31.75 mm