Learn More
Research Products International Corp Direct Thermal Cryo-Spot, 3/8 inch diameter, White, 4000 per roll
SDP
Catalog No. 501978024
Encompass
Polypropylene labels are ideal for use in cryogenic, low temperature and ultra-low temperature freezer storage applications. DirectThermal Cryo-Tags accept printing from Dymo Label Writer and other popular direct thermal printers. Superior labels are chemical resistant and temperature resistant from -196°C to 70°C; freezable in liquid and vapor phase nitrogen. Adhere to most plastics, glass and metals without cracking, peeling or degrading, scuffing or smearing.
Cryogenic, low temperature labels
Temperature resistant -196°C to +70°C
Roll format
Catalog No. 50-197-8024 Supplier Research Products International Corp Supplier No. 249403
Polypropylene labels are ideal for use in cryogenic, low temperature and ultra-low temperature freezer storage applications. DirectThermal Cryo-Tags accept printing from Dymo Label Writer and other popular direct thermal printers. Superior labels are chemical resistant and temperature resistant from -196°C to 70°C; freezable in liquid and vapor phase nitrogen. Adhere to most plastics, glass and metals without cracking, peeling or degrading, scuffing or smearing.
Cryogenic, low temperature labels
Temperature resistant -196°C to +70°C
Roll format