Learn More
MSC DIRECT VENT SAFETY GOGGLE
Catalog No. 5000769387
Encompass
These safety goggles provide reliable eye protection with clear, uncoated polycarbonate lenses. Designed for impact protection, they feature direct ventilation and can be worn over most prescription eyewear, offering enhanced peripheral vision. They meet ANSI Z87.1+ and CSA Z94.3 standards, making them suitable for various industrial and occupational uses.
  • Polycarbonate lens absorbs 99.9% of UV rays
  • Direct ventilation system reduces fog build-up
  • Accommodates most prescription eyewear
  • Adjustable elastic strap provides a secure fit
  • Impact-rated protection meets ANSI Z87.1+ and CSA Z94.3
  • Universal sizing for broad compatibility
  • Offers distinct side-to-side peripheral vision
Catalog No. 50-007-69387 Supplier MSC Supplier No. 59427658
These safety goggles provide reliable eye protection with clear, uncoated polycarbonate lenses. Designed for impact protection, they feature direct ventilation and can be worn over most prescription eyewear, offering enhanced peripheral vision. They meet ANSI Z87.1+ and CSA Z94.3 standards, making them suitable for various industrial and occupational uses.
  • Polycarbonate lens absorbs 99.9% of UV rays
  • Direct ventilation system reduces fog build-up
  • Accommodates most prescription eyewear
  • Adjustable elastic strap provides a secure fit
  • Impact-rated protection meets ANSI Z87.1+ and CSA Z94.3
  • Universal sizing for broad compatibility
  • Offers distinct side-to-side peripheral vision