Learn More
Zymo Research Corporation Direct-zol-96 MagBead RNA (96 Prep)
SDP
Catalog No. NC1822177
Encompass_Preferred
The Direct-zol-96 MagBead RNA kit provides a high-throughput (96-well), magnetic bead-based isolation of high-quality RNA directly from samples in TRI Reagent and similar. The extraction method inactivates viruses and other infectious agents. Total RNA including small/microRNAs (≥17 nt) are effectively isolated from a variety of sample sources (cells, tissues, biological liquids, etc.). The procedure is easy: simply add ethanol and MagBinding Beads to a sample in TRI Reagent, wash and elute the RNA. No phase separation, precipitation, or post-purification steps are necessary. The result is broad range purification of small and large RNAs suitable for subsequent RNA-based methods including Next-Gen sequencing, RT-PCR, transcription profiling, hybridization, etc.
Catalog No. NC1822177 Supplier Zymo Research Corporation Supplier No. R2100
The Direct-zol-96 MagBead RNA kit provides a high-throughput (96-well), magnetic bead-based isolation of high-quality RNA directly from samples in TRI Reagent and similar. The extraction method inactivates viruses and other infectious agents. Total RNA including small/microRNAs (≥17 nt) are effectively isolated from a variety of sample sources (cells, tissues, biological liquids, etc.). The procedure is easy: simply add ethanol and MagBinding Beads to a sample in TRI Reagent, wash and elute the RNA. No phase separation, precipitation, or post-purification steps are necessary. The result is broad range purification of small and large RNAs suitable for subsequent RNA-based methods including Next-Gen sequencing, RT-PCR, transcription profiling, hybridization, etc.