Learn More
Zymo Research Corporation Direct-zol-96 MagBead RNA w/ TRI Reagent (96 preps)
SDP
Catalog No. 501251725
Encompass_Preferred
The extraction method inactivates viruses and other infectious agents. Total RNA (including small/microRNAs) are effectively isolated from a variety of sample sources (cells, tissues, biological liquids, DNA/RNA Shield™ samples, etc.). The procedure is easy: simply add ethanol and MagBinding Beads to a sample in TRIzol/TRI Reagent, wash and elute the RNA. No chloroform, phase separation or precipitation steps are necessary. High-quality total RNA is ready for Next-Gen sequencing, RT-qPCR, transcription profiling, hybridization, etc
Catalog No. 50-125-1725 Supplier Zymo Research Corporation Supplier No. R2101
The extraction method inactivates viruses and other infectious agents. Total RNA (including small/microRNAs) are effectively isolated from a variety of sample sources (cells, tissues, biological liquids, DNA/RNA Shield™ samples, etc.). The procedure is easy: simply add ethanol and MagBinding Beads to a sample in TRIzol/TRI Reagent, wash and elute the RNA. No chloroform, phase separation or precipitation steps are necessary. High-quality total RNA is ready for Next-Gen sequencing, RT-qPCR, transcription profiling, hybridization, etc