Learn More
Zymo Research Corporation Direct-zol DNA Wash 1 (concentrate), 40 ml
SDP
Catalog No. 502037257
Encompass_Preferred
Specifications:
Add 15.5 ml 100% ethanol (16 ml 95% ethanol) to the 24 ml Direct-zol™ DNA Wash 1 concentrate or 61 ml 100% ethanol (64 ml 95% ethanol) to the 96 ml Direct-zol™ DNA Wash 1 concentrate
Catalog No. 50-203-7257 Supplier Zymo Research Corporation Supplier No. R2080240
Specifications:
Add 15.5 ml 100% ethanol (16 ml 95% ethanol) to the 24 ml Direct-zol™ DNA Wash 1 concentrate or 61 ml 100% ethanol (64 ml 95% ethanol) to the 96 ml Direct-zol™ DNA Wash 1 concentrate