Learn More
Zymo Research Corporation Direct-zol™ RNA MicroPrep (200 Preps) w/ Zymo-Spin™ IC Columns (Capped) (Product Supplied w/ 200 ml TRI Reagent™) [Includes R2052 x 1, R2050-1-200 x 1 - packaged separately]
SDP
Catalog No. 501251718
Encompass_Preferred
Simply add ethanol to a TRI Reagent sample, bind directly to the Zymo-Spin™ Column, wash, and elute RNA. No phase separation, precipitation, or post-purification steps are necessary.RNA is high-quality and ready for Next-Gen Sequencing, RTqPCR, transcription profiling, hybridization, etc.Features:1. Efficient recovery of DNA-free Total RNAHigh-quality DNA-free RNA is purified from human epithelial cells using the Direct-zol™ procedure compared to a preparation from Supplier Q.2.Complete miRNA recoveryMicroRNA isolation can use Direct-zol™ RNA kits.The data show RNA purified from TRIzolsamples using the Direct-zol™ RNA MiniPrep compared to a method known to be unbiased2 (mirVana™, Ambion).MicroRNA analysis was performed using miRNAseq (MiSeq, Illumina) and direct hybridization assay (nCounter, Nanostring).
Catalog No. 50-125-1718 Supplier Zymo Research Corporation Supplier No. R2063
Simply add ethanol to a TRI Reagent sample, bind directly to the Zymo-Spin™ Column, wash, and elute RNA. No phase separation, precipitation, or post-purification steps are necessary.RNA is high-quality and ready for Next-Gen Sequencing, RTqPCR, transcription profiling, hybridization, etc.Features:1. Efficient recovery of DNA-free Total RNAHigh-quality DNA-free RNA is purified from human epithelial cells using the Direct-zol™ procedure compared to a preparation from Supplier Q.2.Complete miRNA recoveryMicroRNA isolation can use Direct-zol™ RNA kits.The data show RNA purified from TRIzolsamples using the Direct-zol™ RNA MiniPrep compared to a method known to be unbiased2 (mirVana™, Ambion).MicroRNA analysis was performed using miRNAseq (MiSeq, Illumina) and direct hybridization assay (nCounter, Nanostring).