Learn More
Zymo Research Corporation Direct-zol-96 RNA (4x96 Preps)
SDP
Catalog No. 50444632
Encompass_Preferred
by using this product, total RNA, including small RNAs (17-200 nt), is effectively isolated from a variety ofsample sources (cells, tissues, serum, plasma, blood, biological liquids, etc.)The extraction method inactivates viruses and other infectious agents.The procedure is easy: simply apply a sample in TRI Reagent to the Zymo-Spin™ I-96 Plate, then bind, wash, and elute the RNA. No phase separation, precipitation, or post-purification steps are necessary. The result is high-quality RNA suitable for subsequent RNA-based methods including Next-Gen sequencing, RT-qPCR, hybridization, etc.
Catalog No. 50-444-632 Supplier Zymo Research Corporation Supplier No. R2056
by using this product, total RNA, including small RNAs (17-200 nt), is effectively isolated from a variety ofsample sources (cells, tissues, serum, plasma, blood, biological liquids, etc.)The extraction method inactivates viruses and other infectious agents.The procedure is easy: simply apply a sample in TRI Reagent to the Zymo-Spin™ I-96 Plate, then bind, wash, and elute the RNA. No phase separation, precipitation, or post-purification steps are necessary. The result is high-quality RNA suitable for subsequent RNA-based methods including Next-Gen sequencing, RT-qPCR, hybridization, etc.