Learn More
Sigma Aldrich Fine Chemicals Biosciences DirectLoadTM 1 kb DNA Ladd
Catalog No. 502901458
Encompass
Sigmas DirectLoad(TM) 1 kb Ladder contains 11 fragments consisting of 500 bp repeats from 0.5 to 3 kb 1 kb repeats from 3 to 6 kb and 2 kb repeats from 6 to 10 kb. for NA electrophoresis 5 ul of the marker can be loaded directly into a single lane on an agarose or polyacrylamide gel. Suitable for use in Northern and Southern blotting.
Catalog No. 50-290-1458 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D39371VL
Sigmas DirectLoad(TM) 1 kb Ladder contains 11 fragments consisting of 500 bp repeats from 0.5 to 3 kb 1 kb repeats from 3 to 6 kb and 2 kb repeats from 6 to 10 kb. for NA electrophoresis 5 ul of the marker can be loaded directly into a single lane on an agarose or polyacrylamide gel. Suitable for use in Northern and Southern blotting.