Learn More
Sigma Aldrich Fine Chemicals Biosciences DirectLoadTM PCR 100 bp Lo
Catalog No. 502900709
Encompass
Sigmas DirectLoad(TM) PCR 100 bp Low Ladder contains 10 dsDNA fragments from 100 - 1 000 bases evenly spaced in 100 bp repeats. Ladder should be loaded in a single lane on an agarose gel.
Catalog No. 50-290-0709 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D36871VL
Sigmas DirectLoad(TM) PCR 100 bp Low Ladder contains 10 dsDNA fragments from 100 - 1 000 bases evenly spaced in 100 bp repeats. Ladder should be loaded in a single lane on an agarose gel.