Learn More
INTACT GENOMICS INC DirectPlate 10B Chemically Competent Cells 12x50µl
Catalog No. 5000083073
Encompass
Intact Genomics (ig) DirectPlate™ Competent cells offer simple, fast, and robust results for your DNA transformation needs. It's a perfect choice for researchers looking to simplify their transformation workflow by eliminating heat shock, lengthy incubations, and time-consuming outgrowth procedures. Simply mix and directly plate! DirectPlate™ 10B chemically competent E. coli cells provide higher transformation efficiency than any competitor’s similar product and are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning. Our DirectPlate™ 10B chemically competent cells have the highest transformation efficiency on the market.
Catalog No. 50-000-83073 Supplier INTACT GENOMICS INC Supplier No. 101512
Intact Genomics (ig) DirectPlate™ Competent cells offer simple, fast, and robust results for your DNA transformation needs. It's a perfect choice for researchers looking to simplify their transformation workflow by eliminating heat shock, lengthy incubations, and time-consuming outgrowth procedures. Simply mix and directly plate! DirectPlate™ 10B chemically competent E. coli cells provide higher transformation efficiency than any competitor’s similar product and are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning. Our DirectPlate™ 10B chemically competent cells have the highest transformation efficiency on the market.