Learn More
INTACT GENOMICS INC DirectPlate 5-alpha Chemically Competent Cells 36x50l
Catalog No. 5000082884
Encompass
Intact Genomics (ig) DirectPlate™ Competent cells offer simple, fast, and robust results for your DNA transformation needs. It's a perfect choice for researchers looking to simplify their transformation workflow by eliminating heat shock, lengthy incubations, and time-consuming outgrowth procedures. Simply mix and directly plate! DirectPlate™ 5-Alpha chemically competent E. coli cells provide higher transformation efficiency than any competitors similar product and are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning.
Catalog No. 50-000-82884 Supplier INTACT GENOMICS INC Supplier No. 101336
Intact Genomics (ig) DirectPlate™ Competent cells offer simple, fast, and robust results for your DNA transformation needs. It's a perfect choice for researchers looking to simplify their transformation workflow by eliminating heat shock, lengthy incubations, and time-consuming outgrowth procedures. Simply mix and directly plate! DirectPlate™ 5-Alpha chemically competent E. coli cells provide higher transformation efficiency than any competitors similar product and are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning.