Learn More
INTACT GENOMICS INC DirectPlate TG1 Phage Display Chemically Competent Cells
Supplier:  INTACT GENOMICS INC 102412
Encompass
Intact Genomics (ig) DirectPlate™ Competent cells offer simple, fast, and robust results for your DNA transformation needs. DirectPlate™ TG1 Phage Display chemically competent E. coli cells are a perfect choice for researchers looking to simplify their transformation workflow by eliminating heat shock, lengthy incubations, and time-consuming outgrowth procedures. Simply mix and directly plate! DirectPlate™ TG1 Phage Display chemically competent E. coli cells provide higher transformation efficiency than any competitor’s similar product and are suitable for high-efficiency transformation in a wide variety of applications such as cloning, phage display libraries and sub-cloning.Product Includes and Storage:DirectPlate™ TG1 Phage Display competent cells: -80 ºCpUC19 control DNA: -20 ºC
Catalog No. 50-000-82889