Learn More
INTACT GENOMICS INC DirectPlate XL™ DH10B Chemically Competent Cells 36x50µl
Catalog No. 5000082979
Encompass
Intact Genomics (ig) DirectPlate-XL™ DH10B Competent cells offer simple, fast, and robust results for your large DNA transformation needs – directly clone large DNA up to >250kb! DirectPlate-XL™ DH10B chemically competent E. coli cells are a perfect choice for researchers looking to simplify their large DNA transformation workflow by eliminating heat shock, lengthy incubations, and time-consuming outgrowth procedures. Simply mix and directly plate! DirectPlate-XL™ DH10B chemically competent E. coli cells provide higher transformation efficiency than any competitor’s similar product and are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning. Using proprietary technology, these cells can be used for the transformation of extremely large DNA (i.e. >10kb up to >250kb).Product Includes and Storage:DirectPlate-XL™ DH10B competent cells: -80 ºCpUC19 control DNA: -20 ºC
Catalog No. 50-000-82979 Supplier INTACT GENOMICS INC Supplier No. 109436
Intact Genomics (ig) DirectPlate-XL™ DH10B Competent cells offer simple, fast, and robust results for your large DNA transformation needs – directly clone large DNA up to >250kb! DirectPlate-XL™ DH10B chemically competent E. coli cells are a perfect choice for researchers looking to simplify their large DNA transformation workflow by eliminating heat shock, lengthy incubations, and time-consuming outgrowth procedures. Simply mix and directly plate! DirectPlate-XL™ DH10B chemically competent E. coli cells provide higher transformation efficiency than any competitor’s similar product and are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning. Using proprietary technology, these cells can be used for the transformation of extremely large DNA (i.e. >10kb up to >250kb).Product Includes and Storage:DirectPlate-XL™ DH10B competent cells: -80 ºCpUC19 control DNA: -20 ºC