Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dirhodium tetraacetate | 15956-28-2 | 441.99 | 25MG
SDP

Catalog No. 5000449387
Encompass_Preferred

Dirhodium tetraacetate is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Biochemical reagent for life science related research
  • Solid appearance
  • Light green to green color
  • Soluble in DMSO
  • Shipped at room temperature
  • Store at 4°C, protect from light, stored under nitrogen

Catalog No. 50-004-49387 Supplier Medchemexpress LLC Supplier No. 25MGHYW001951
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dirhodium tetraacetate is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Biochemical reagent for life science related research
  • Solid appearance
  • Light green to green color
  • Soluble in DMSO
  • Shipped at room temperature
  • Store at 4°C, protect from light, stored under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.