Learn More
Selleck Chemical LLC Dirithromycin S4213-50mg
SDP
Catalog No. 503108545
Encompass_Preferred
Dirithromycin (LY-237216 ASE 136 Dynabac) is a macrolide glycopeptide antibiotic by binding to the 50S subunit of the 70S bacterial ribosome to inhibit the translocation of peptides
Catalog No. 50-310-8545 Supplier Selleck Chemical LLC Supplier No. S421350MG
Dirithromycin (LY-237216 ASE 136 Dynabac) is a macrolide glycopeptide antibiotic by binding to the 50S subunit of the 70S bacterial ribosome to inhibit the translocation of peptides