Learn More
Cayman Chemical DIrIthromycIn 1g
SDP
Catalog No. 502981031
Encompass_Preferred
A macrolide antibiotic and prodrug of erythromycylamine that has outstanding activity against Campylobacter
Catalog No. 50-298-1031 Supplier Cayman Chemical Supplier No. 194661 G
A macrolide antibiotic and prodrug of erythromycylamine that has outstanding activity against Campylobacter