Learn More
Apexbio Technology LLC Dirithromycin 62013-04-1 10mM (in 1mL Ethanol)
SDP
Catalog No. 5000061972
Encompass_Preferred
Dirithromycin is a macrolide glycopeptide antibiotic which exerts antibacterial activity by targeting the bacterial ribosomal 50S subunit thereby inhibiting peptide chain translocation during protein synthesis It effectively interferes with bacterial growth and proliferation by restricting the translation process In microbiological and pharmacological research Dirithromycin is utilized as a tool compound to investigate ribosomal function antibiotic resistance mechanisms and bacterial protein synthesis pathways in vitro In susceptibility assays against various bacteria Dirithromycin demonstrates inhibition of ribosome-mediated functions with reported IC50 values typically ranging from 0 1 to 2 g/mL depending on bacterial species and experimental conditions
Catalog No. 50-000-61972 Supplier Apexbio Technology LLC Supplier No. B17315.1
Dirithromycin is a macrolide glycopeptide antibiotic which exerts antibacterial activity by targeting the bacterial ribosomal 50S subunit thereby inhibiting peptide chain translocation during protein synthesis It effectively interferes with bacterial growth and proliferation by restricting the translation process In microbiological and pharmacological research Dirithromycin is utilized as a tool compound to investigate ribosomal function antibiotic resistance mechanisms and bacterial protein synthesis pathways in vitro In susceptibility assays against various bacteria Dirithromycin demonstrates inhibition of ribosome-mediated functions with reported IC50 values typically ranging from 0 1 to 2 g/mL depending on bacterial species and experimental conditions