Learn More
Cayman Chemical DIrIthromycIn 500mg
SDP
Catalog No. 502985438
Encompass_Preferred
A macrolide antibiotic and prodrug of erythromycylamine that has outstanding activity against Campylobacter
Catalog No. 50-298-5438 Supplier Cayman Chemical Supplier No. 19466500 MG
A macrolide antibiotic and prodrug of erythromycylamine that has outstanding activity against Campylobacter