Learn More
Medchemexpress LLC HY-B0643 500mg Medchemexpress, Dirithromycin CAS:62013-04-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0643500MG
Encompass_Preferred
Medchemexpress, HY-B0643 500mg Dirithromycin CAS:62013-04-1 Dirithromycin (LY237216), a derivative of Erythromycin, is a potent and orally active semi-synthetic macrolide antibiotic. Dirithromycin is active against gram-positive bacteria, Legionella spp., Helicobacter pylori, and Chlamydia trachomatis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2629