Learn More
Apexbio Technology LLC Dirithromycin, 50mg. Cas: 62013-04-1 MFCD: MFCD00865041. Macrolide glycopeptide antibiotic
SDP
Catalog No. 501905280
Encompass_Preferred
Dirithromycin is a macrolide glycopeptide antibiotic by binding to the 50S subunit of the 70S bacterial ribosome to inhibit the translocation of peptides. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5280 Supplier Apexbio Technology LLC Supplier No. B173150
Dirithromycin is a macrolide glycopeptide antibiotic by binding to the 50S subunit of the 70S bacterial ribosome to inhibit the translocation of peptides. Other sizes are available. Please inqury us for quote.