Learn More
Medchemexpress LLC Dirithromycin | 62013-04-1 | 98.0% | 1 ML
SDP
Catalog No. 5000409048
Encompass_Preferred
Dirithromycin is a potent, orally active, semi-synthetic macrolide antibiotic derived from Erythromycin. It demonstrates activity against gram-positive bacteria, Legionella spp., Helicobacter pylori, and Chlamydia trachomatis. This product is intended for research use only.
  • Semi-synthetic macrolide antibiotic
  • Derived from Erythromycin
  • Active against gram-positive bacteria
  • Effective against *Legionella spp.*, *Helicobacter pylori*, and *Chlamydia trachomatis*
  • Demonstrates potent *in vitro* activity against *Legionella* and *Helicobacter pylori*
  • Effective in experimental infections caused by *S. aureus*, *S. pyogenes*, and *S. pneumoniae*
Catalog No. 50-004-09048 Supplier Medchemexpress LLC Supplier No. HYB064310MM/1ML
Dirithromycin is a potent, orally active, semi-synthetic macrolide antibiotic derived from Erythromycin. It demonstrates activity against gram-positive bacteria, Legionella spp., Helicobacter pylori, and Chlamydia trachomatis. This product is intended for research use only.
  • Semi-synthetic macrolide antibiotic
  • Derived from Erythromycin
  • Active against gram-positive bacteria
  • Effective against *Legionella spp.*, *Helicobacter pylori*, and *Chlamydia trachomatis*
  • Demonstrates potent *in vitro* activity against *Legionella* and *Helicobacter pylori*
  • Effective in experimental infections caused by *S. aureus*, *S. pyogenes*, and *S. pneumoniae*