Learn More
Medchemexpress LLC Dirithromycin | 62013-04-1 | 98.0% | 5 G
SDP
Catalog No. 5000407714
Encompass_Preferred
Dirithromycin is a potent and orally active semi-synthetic macrolide antibiotic, derived from Erythromycin. It is active against gram-positive bacteria, *Legionella* spp., *Helicobacter pylori*, and *Chlamydia trachomatis*. Intended for research use only.
  • Exhibits antimicrobial activity similar to Erythromycin
  • Shows excellent activity against several strains of *Legionella*
  • Demonstrates potent activity against several strains of *Helicobacter pylori*
  • Effective against experimental infections caused by *S. aureus*, *S. pyogenes*, and *S. pneumoniae* in mice
Catalog No. 50-004-07714 Supplier Medchemexpress LLC Supplier No. HYB06435G
Dirithromycin is a potent and orally active semi-synthetic macrolide antibiotic, derived from Erythromycin. It is active against gram-positive bacteria, *Legionella* spp., *Helicobacter pylori*, and *Chlamydia trachomatis*. Intended for research use only.
  • Exhibits antimicrobial activity similar to Erythromycin
  • Shows excellent activity against several strains of *Legionella*
  • Demonstrates potent activity against several strains of *Helicobacter pylori*
  • Effective against experimental infections caused by *S. aureus*, *S. pyogenes*, and *S. pneumoniae* in mice