Learn More
Medchemexpress LLC HY-U00070 1mg Medchemexpress, Dirlotapide CAS:481658-94-0 Purity:>98%
SDP
Catalog No. 502083186
Encompass_Preferred
Medchemexpress, HY-U00070 1mg Dirlotapide CAS:481658-94-0 Dirlotapide (CP742033) is a gut-selective inhibitor of microsomal triglyceride transfer protein (MTP) that reliably produces weight loss in obese dogs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3186 Supplier Medchemexpress LLC Supplier No. HYU000701MG
Medchemexpress, HY-U00070 1mg Dirlotapide CAS:481658-94-0 Dirlotapide (CP742033) is a gut-selective inhibitor of microsomal triglyceride transfer protein (MTP) that reliably produces weight loss in obese dogs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.