Learn More
Med Vet International Zoetis Diroban Sterile Powder For Injection, 2mL, 5/Pack
Catalog No. 502827827
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Diroban (melarsomine dihydrochloride) Sterile Powder for Injection is indicated for the treatment of stabilized Class 1, 2, and 3 heartworm disease caused by immature (4 month-old, stage L5) to mature adult infections of Dirofilaria immitis in dogs. See package insert for complete product details.
Catalog No. 50-282-7827 Supplier Med Vet International Supplier No. RXDIROBAN5
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Diroban (melarsomine dihydrochloride) Sterile Powder for Injection is indicated for the treatment of stabilized Class 1, 2, and 3 heartworm disease caused by immature (4 month-old, stage L5) to mature adult infections of Dirofilaria immitis in dogs. See package insert for complete product details.