Learn More
Cayman Chemical DIroxImel Fumarate 10mg
SDP
Catalog No. 502939658
Encompass_Preferred
A prodrug form of monomethyl fumarate; undergoes esterase cleavage in the gut to release monomethyl fumarate
Catalog No. 50-293-9658 Supplier Cayman Chemical Supplier No. 2911110 MG
A prodrug form of monomethyl fumarate; undergoes esterase cleavage in the gut to release monomethyl fumarate