Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diroximel fumarate | 1577222-14-0 | ≥98.0% | 255.22 g/mol | C11H13NO6 | 10 MG
SDP

Catalog No. 5000214219
Encompass_Preferred

Diroximel fumarate is an orally active prodrug of monomethyl fumarate used as a research reagent to study Nrf2 activation and pathways relevant to multiple sclerosis. Supplied as a powder in small quantities for in vitro and preclinical use, it should be handled and stored according to the supplier's safety data sheet and product datasheet.

  • Orally active monomethyl fumarate prodrug for Nrf2 pathway research.
  • Supplied as a powder suitable for in vitro and preclinical studies.
  • Available in small package sizes appropriate for screening and pilot experiments.
  • Molecular formula C11H13NO6; molecular weight 255.22 g/mol.
  • Store powder at -20°C for long-term stability or 4°C for shorter storage.
  • CAS number 1577222-14-0 for unambiguous identification.

Catalog No. 50-002-14219 Supplier Medchemexpress LLC Supplier No. HY10037510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diroximel fumarate is an orally active prodrug of monomethyl fumarate used as a research reagent to study Nrf2 activation and pathways relevant to multiple sclerosis. Supplied as a powder in small quantities for in vitro and preclinical use, it should be handled and stored according to the supplier's safety data sheet and product datasheet.

  • Orally active monomethyl fumarate prodrug for Nrf2 pathway research.
  • Supplied as a powder suitable for in vitro and preclinical studies.
  • Available in small package sizes appropriate for screening and pilot experiments.
  • Molecular formula C11H13NO6; molecular weight 255.22 g/mol.
  • Store powder at -20°C for long-term stability or 4°C for shorter storage.
  • CAS number 1577222-14-0 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.