Learn More
Medchemexpress LLC Diroximel fumarate | 1577222-14-0 | 99.9% | 255.22 g/mol | C11H13NO6 | 1 ML
SDP
Catalog No. 5000214290
Encompass_Preferred
Diroximel fumarate is an orally active prodrug of monomethyl fumarate used in preclinical research to probe Nrf2 pathway modulation and multiple sclerosis-related biology. Supplied as a ready-to-use solution, it is intended for laboratory research use only and not for human or clinical applications.
  • Ready-to-use 10 mM solution in DMSO for immediate experimental use.
  • High purity (99.90%) suitable for biochemical and cellular assays.
  • Molecular weight 255.22 g/mol, molecular formula C11H13NO6.
  • Prodrug of monomethyl fumarate for Nrf2 pathway and MS-related research.
  • Provided in a 1 mL volume for small-scale assays and dose-response studies.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-14290 Supplier Medchemexpress LLC Supplier No. HY10037510MM 1ML
Diroximel fumarate is an orally active prodrug of monomethyl fumarate used in preclinical research to probe Nrf2 pathway modulation and multiple sclerosis-related biology. Supplied as a ready-to-use solution, it is intended for laboratory research use only and not for human or clinical applications.
  • Ready-to-use 10 mM solution in DMSO for immediate experimental use.
  • High purity (99.90%) suitable for biochemical and cellular assays.
  • Molecular weight 255.22 g/mol, molecular formula C11H13NO6.
  • Prodrug of monomethyl fumarate for Nrf2 pathway and MS-related research.
  • Provided in a 1 mL volume for small-scale assays and dose-response studies.
  • For research use only; not for human or clinical use.