Learn More
Medchemexpress LLC Diroximel fumarate | 1577222-14-0 | MFCD10057039 | >98.0% | 255.22 | C11H13NO6 | 25 MG
SDP
Catalog No. 5000214554
Encompass_Preferred
Diroximel fumarate is an orally active prodrug of monomethyl fumarate used in research to investigate Nrf2 pathway activation and multiple sclerosis-related pharmacology. The compound is provided with analytical data for identification and formulation of stock solutions.
  • Orally active prodrug of monomethyl fumarate.
  • Useful for in vitro and in vivo studies of Nrf2-mediated pathways.
  • Provided with analytical specifications including CAS, formula, and molecular weight for traceability.
  • Available in mg-scale quantities suitable for preclinical research.
Catalog No. 50-002-14554 Supplier Medchemexpress LLC Supplier No. HY10037525MG
Diroximel fumarate is an orally active prodrug of monomethyl fumarate used in research to investigate Nrf2 pathway activation and multiple sclerosis-related pharmacology. The compound is provided with analytical data for identification and formulation of stock solutions.
  • Orally active prodrug of monomethyl fumarate.
  • Useful for in vitro and in vivo studies of Nrf2-mediated pathways.
  • Provided with analytical specifications including CAS, formula, and molecular weight for traceability.
  • Available in mg-scale quantities suitable for preclinical research.