Learn More
TARGETMOL CHEMICALS INC Diroximel fumarate 25MG
SDP
Catalog No. 502878767
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Diroximel fumarate (Diroximel Fumarete), a prodrug of monomethyl fumarate in a controlled-release formulation, rapidly and potently converts to MMF in the body. Purity 99.88%
Catalog No. 50-287-8767 Supplier TARGETMOL CHEMICALS INC Supplier No. T427125MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Diroximel fumarate (Diroximel Fumarete), a prodrug of monomethyl fumarate in a controlled-release formulation, rapidly and potently converts to MMF in the body. Purity 99.88%