Learn More
Cayman Chemical Diroximel Fumarate, Purity: ≥98%, CAS: 1577222-14-0, 50 mg
SDP
Catalog No. 502939692
Encompass_Preferred
A prodrug form of monomethyl fumarate; undergoes esterase cleavage in the gut to release monomethyl fumarate
Catalog No. 50-293-9692 Supplier Cayman Chemical Supplier No. 2911150 MG
A prodrug form of monomethyl fumarate; undergoes esterase cleavage in the gut to release monomethyl fumarate