Learn More
Selleck Chemical LLC Diroximel Fumarate
SDP
Catalog No. 5000002990
Encompass_Preferred
Diroximel Fumarate (ALKS-8700) is a prodrug of monomethyl fumarate in a controlled-release formulation that rapidly and efficiently converts to MMF in the body
Catalog No. 50-000-02990 Supplier Selleck Chemical LLC Supplier No. S64215MG
Diroximel Fumarate (ALKS-8700) is a prodrug of monomethyl fumarate in a controlled-release formulation that rapidly and efficiently converts to MMF in the body