Learn More
Chemglass Life Sciences Distilling Receiver, 50mL, 24/40, Complete, Approx 413mm Height x 160mm Width
SDP
Catalog No. CG127901
Encompass_Preferred
Distilling Receiver, 50mL, 24/40, Complete, Approx 413mm Height x 160mm Width. Radial style distillation receiver allows four flasks to be progressively filled. Upper vacuum adapter is bent at a 105º angle. Complete item consists of a vacuum adapter, distribution adapter, four of the listed size flasks with hooks and ten CG-110 stainless steel springs.
Catalog No. CG127901 Supplier Chemglass Life Sciences Supplier No. CG127901
Distilling Receiver, 50mL, 24/40, Complete, Approx 413mm Height x 160mm Width. Radial style distillation receiver allows four flasks to be progressively filled. Upper vacuum adapter is bent at a 105º angle. Complete item consists of a vacuum adapter, distribution adapter, four of the listed size flasks with hooks and ten CG-110 stainless steel springs.