Learn More
Aviva Systems Biology Corp DIS3 Antibody - N-terminal region (ARP40793_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40793_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DIS3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DIS3. Alternate names: DKFZp667L1817, FLJ10484, KIAA1008, MGC33035, RP11-342J4.3, RRP44, bA555G22.1, dis3p, EXOSC11ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57625