Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Disappearing Rainbow
GREENER_CHOICESCI_ED

Catalog No. S98653 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S98653 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Phenolphthalein/Methanol/Methyl isobutyl ketone, which are known to the State of California to cause cancer, birth defects and reproductive harm.
Catalog No. S98653 Supplier Innovating Science™ Supplier No. IS7039
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Phenolphthalein/Methanol/Methyl isobutyl ketone, which are known to the State of California to cause cancer, birth defects and reproductive harm.

Includes

Materials for five demonstrations: 800 mL Sodium Hydroxide solution; 50 mL Sodium Hydroxide, 3M; 1 L Hydrochloric Acid, 0.01M; violet, blue, green, yellow, orange and red indicators; and instructions

Use acids and bases with three indicators to create a rainbow of colors.

  • In a basic solution, phenolphtalein is red, p-nitrophenol is yellow and thymolphthalein is blue
  • These can be combined in various ways to create all the colors of the rainbow
  • If done correctly, students only see the acid or base being poured, adding to the impressive display

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Use acids and bases with three indicators to create a rainbow of colors
Includes 25 mL Red indicator UN1170, 25 mL orange indicator UN1170, 25 mL yellow indicator UN1170, 25 mL green indicator UN1170, 25 mL blue indicator UN1170, 25 mL violet indicator UN1170, 2 x 30 mL Hydrochloric Acid 1N UN1789, 2 x 30 mL Sodium Hydroxide 1N UN1824, 30 mL Sodium Hydroxide Sol 6M (6N) UN1824
Quantity 1
Dimensions (L x W x H) 5.5 x 4.25 x 6.75 in.
Class Size 6 Lab Groups
Product Type Disappearing Rainbow Demonstration Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.