Learn More
Waters Corp Rupture Disk, 10,000 PSI, 1/4 in., 72°F Rupture
Catalog No. NC3346497 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rupture disk is designed for use in high-pressure CO2 pump systems. It is specified for a pressure of 10,000 PSI and has a 1/4 in. connection, with a rupture temperature of 72°F.
  • Specified for 10,000 PSI pressure
  • Features a 1/4 in. connection
  • Designed to rupture at 72°F
  • Suitable for CO2 pump systems
Catalog No. NC3346497 Supplier Waters Corp Supplier No. 700006552
This rupture disk is designed for use in high-pressure CO2 pump systems. It is specified for a pressure of 10,000 PSI and has a 1/4 in. connection, with a rupture temperature of 72°F.
  • Specified for 10,000 PSI pressure
  • Features a 1/4 in. connection
  • Designed to rupture at 72°F
  • Suitable for CO2 pump systems