Learn More
MSC Clarke Walk Behind Disc Floor Scrubber, 28 inch, 23 Gallon, Battery
Catalog No. 502836406
Encompass
The Clarke Focus II Disc 28 Walk Behind Floor Scrubber is an automatic cleaning machine designed for efficient floor maintenance in various commercial and industrial settings. It lays down cleaning solution, scrubs, and then vacuums away dirty solution, leaving floors clean and dry in one pass.
  • 28 inch cleaning path
  • Disc design for deep cleaning
  • Intuitive one-touch controls
  • Cordless operation with lead acid/wet batteries
  • Quiet operation at 65 decibels
  • Two heavy-duty brush motors, 80 to 150 lb. pressure
  • Large 23 gallon solution and recovery tanks
  • Electronic float switch for full recovery tank
  • 35 inch squeegee for efficient solution pickup
  • Security key switch, safety palm switches, and thermal overload protection
  • Onboard battery charger
  • Easy to maneuver with adjustable squeegee
Catalog No. 50-283-6406 Supplier MSC Supplier No. 35706845
Add to Cart
Add to Cart
Request a Quote
The Clarke Focus II Disc 28 Walk Behind Floor Scrubber is an automatic cleaning machine designed for efficient floor maintenance in various commercial and industrial settings. It lays down cleaning solution, scrubs, and then vacuums away dirty solution, leaving floors clean and dry in one pass.
  • 28 inch cleaning path
  • Disc design for deep cleaning
  • Intuitive one-touch controls
  • Cordless operation with lead acid/wet batteries
  • Quiet operation at 65 decibels
  • Two heavy-duty brush motors, 80 to 150 lb. pressure
  • Large 23 gallon solution and recovery tanks
  • Electronic float switch for full recovery tank
  • 35 inch squeegee for efficient solution pickup
  • Security key switch, safety palm switches, and thermal overload protection
  • Onboard battery charger
  • Easy to maneuver with adjustable squeegee