Learn More
MSC Clarke Walk Behind Floor Scrubber, 26 in Cleaning Path, AGM, Disc
Catalog No. 502836907
Encompass
The Focus II Disc 26 AGM Cordless Walk Behind Floor Scrubber offers a deeper clean for various establishments. It features a cordless design with long-lasting, maintenance-free AGM batteries and an intuitive one-touch control panel.
  • 26 inch cleaning path
  • Chemical mixing system
  • Cordless design with AGM batteries
  • Intuitive one-touch control panel
  • Battery-operated for flexibility
  • Maintenance-free AGM batteries
  • Disc design with two heavy-duty brush motors
  • Brush applies 80 to 150 lb. pressure
  • Brush speed of 200 RPM
  • Large recovery tank opening
  • Electronic float switch for vacuum motor cut-off
  • 32 1/2 inch squeegee width
  • Safety key switch, palm switches, thermal overload protection
  • Operates at 65 decibels
  • 23 gallon solution and recovery tanks
Catalog No. 50-283-6907 Supplier MSC Supplier No. 35706894
Add to Cart
Add to Cart
Request a Quote
The Focus II Disc 26 AGM Cordless Walk Behind Floor Scrubber offers a deeper clean for various establishments. It features a cordless design with long-lasting, maintenance-free AGM batteries and an intuitive one-touch control panel.
  • 26 inch cleaning path
  • Chemical mixing system
  • Cordless design with AGM batteries
  • Intuitive one-touch control panel
  • Battery-operated for flexibility
  • Maintenance-free AGM batteries
  • Disc design with two heavy-duty brush motors
  • Brush applies 80 to 150 lb. pressure
  • Brush speed of 200 RPM
  • Large recovery tank opening
  • Electronic float switch for vacuum motor cut-off
  • 32 1/2 inch squeegee width
  • Safety key switch, palm switches, thermal overload protection
  • Operates at 65 decibels
  • 23 gallon solution and recovery tanks