Learn More
MSC Nilfisk Walk-Behind Floor Scrubber, Disc 28, 242Ah, Chemical Mixing
Catalog No. 502836402
Encompass
This floor scrubber is designed for efficient, everyday cleaning, powered by two heavy-duty brush motors. It aims to improve floor cleaning efficiency, offering a disc-powered scrub deck suitable for various floor types.
  • Quiet operation for noise-sensitive areas.
  • Optional chemical mixing system for precise chemical and water mixtures.
  • Patented pulse-modulated solution control for consistent solution delivery.
  • Gimble-mounted brushes and pad drivers adapt to uneven floors.
  • Ergonomically placed controls for easy operation.
  • Onboard battery charger reduces downtime.
  • Green-certified equipment.
  • High capacity and long run times.
Catalog No. 50-283-6402 Supplier MSC Supplier No. 35706829
Add to Cart
Add to Cart
Request a Quote
This floor scrubber is designed for efficient, everyday cleaning, powered by two heavy-duty brush motors. It aims to improve floor cleaning efficiency, offering a disc-powered scrub deck suitable for various floor types.
  • Quiet operation for noise-sensitive areas.
  • Optional chemical mixing system for precise chemical and water mixtures.
  • Patented pulse-modulated solution control for consistent solution delivery.
  • Gimble-mounted brushes and pad drivers adapt to uneven floors.
  • Ergonomically placed controls for easy operation.
  • Onboard battery charger reduces downtime.
  • Green-certified equipment.
  • High capacity and long run times.