Learn More
MSC Clarke Floor Scrubber, 26 in Cleaning Path, AGM Battery
Catalog No. 502836407
Encompass
The Focus II Disc Walk Behind Scrubber is designed with a 26 in cleaning path and is powered by an AGM battery. It features a 23 gal solution tank and a 23 gal recovery tank. The durable polyethylene housing ensures longevity, while the 32.5 in squeegee width provides efficient water pick-up. This self-propelled scrubber includes a 0.75 horsepower motor with a brush speed of 200, and is equipped with a deadman switch for enhanced safety.
  • 23 gal solution tank
  • 23 gal recovery tank
  • Polyethylene housing
  • 32.5 in squeegee width
  • Self-propelled operation
  • 0.75 horsepower motor
  • 200 brush speed
  • Deadman switch
Catalog No. 50-283-6407 Supplier MSC Supplier No. 35706878
Add to Cart
Add to Cart
Request a Quote
The Focus II Disc Walk Behind Scrubber is designed with a 26 in cleaning path and is powered by an AGM battery. It features a 23 gal solution tank and a 23 gal recovery tank. The durable polyethylene housing ensures longevity, while the 32.5 in squeegee width provides efficient water pick-up. This self-propelled scrubber includes a 0.75 horsepower motor with a brush speed of 200, and is equipped with a deadman switch for enhanced safety.
  • 23 gal solution tank
  • 23 gal recovery tank
  • Polyethylene housing
  • 32.5 in squeegee width
  • Self-propelled operation
  • 0.75 horsepower motor
  • 200 brush speed
  • Deadman switch